Methyl 3-O-benzyl-β-D-galactopyranoside ≥98%

Supplier: Apollo Scientific

BICL4171-1G BICL4171-500MG
APOSBICL4171-1GEA 1050 EUR
APOSBICL4171-1G APOSBICL4171-500MG
Methyl 3-O-benzyl-β-D-galactopyranoside ≥98%
Methyl 3-O-benzyl-β-D-galactopyranoside
Storage Temperature: Ambient
CAS Number: 81371-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR