Methyl 2,4,6-tri-O-acetyl-3-O-benzyl-β-D-galactopyranoside ≥98%

Supplier: Apollo Scientific

BICL4172-1G BICL4172-500MG BICL4172-250MG
APOSBICL4172-1GEA 1010 EUR
APOSBICL4172-1G APOSBICL4172-500MG APOSBICL4172-250MG
Methyl 2,4,6-tri-O-acetyl-3-O-benzyl-β-D-galactopyranoside ≥98%
Methyl 2,4,6-tri-O-acetyl-3-O-benzyl-β-D-galactopyranoside
Storage Temperature: Ambient
CAS Number: 81348-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR