Naproxen-acyl-β-D-glucuronide ≥98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(6-Methoxynaphthalen-2-yl)propanoic acid-acyl-β-D-glucuronide

BICL4179-1MG BICL4179-2MG
APOSBICL4179-1MGEA 545 EUR
APOSBICL4179-1MG APOSBICL4179-2MG
Naproxen-acyl-β-D-glucuronide ≥98%
Naproxen-acyl-β-D-glucuronide
Formula: C₂₀H₂₂O₉
MW: 406.383 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00869959
CAS Number: 41945-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR