Perindopril-acyl-β-D-glucuronide ≥93%
Supplier: Apollo Scientific
BICL4187-1MG
BICL4187-2MG
APOSBICL4187-1MGEA
517
EUR
APOSBICL4187-1MG
APOSBICL4187-2MG
Perindopril-acyl-β-D-glucuronide ≥93%
Perindopril-acyl-β-D-glucuronide
Storage Temperature:
Ambient |
CAS Number:
120398-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...