1-Deoxy-D-galactitol 98+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
L-Fucitol, NSC 1957, 6-Deoxy-L-galactitol
BICL4223-1G
BICL4223-250MG
BICL4223-500MG
APOSBICL4223-1GEA
866
EUR
APOSBICL4223-1G
APOSBICL4223-250MG
APOSBICL4223-500MG
1-Deoxy-D-galactitol 98+%
1-Deoxy-D-galactitol
Formula:
C₆H₁₄O₅ MW: 166,172 g/mol Melting Pt: 154…155 °C Storage Temperature: Freezer |
MDL Number:
MFCD00083329 CAS Number: 13074-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...