Benzyl β-D-arabinopyranoside ≥98%

Supplier: Apollo Scientific

Synonyms: NSC 2561

BICL4248-1G BICL4248-2G BICL4248-5G
APOSBICL4248-1GEA 351 EUR
APOSBICL4248-1G APOSBICL4248-2G APOSBICL4248-5G
Benzyl β-D-arabinopyranoside ≥98%
Benzyl β-D-arabinopyranoside
Formula: C₁₂H₁₆O₅
MW: 240,252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00225631
CAS Number: 5329-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR