2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl azide ≥97%
Supplier: Apollo Scientific
BICL4249-1G
BICL4249-2G
BICL4249-500MG
APOSBICL4249-1GEA
560
EUR
APOSBICL4249-1G
APOSBICL4249-2G
APOSBICL4249-500MG
2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl azide ≥97%
2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl azide
Formula:
C₁₄H₁₉N₃O₉ MW: 373,315 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05664724 CAS Number: 53784-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...