Methyl β-D-mannopyranoside isopropylate ≥98%

Supplier: Apollo Scientific

Synonyms: Methyl β-D-mannopyranoside 140 molar % isopropanol

BICL4264-100MG BICL4264-250MG BICL4264-500MG
APOSBICL4264-100MGEA 351 EUR
APOSBICL4264-100MG APOSBICL4264-250MG APOSBICL4264-500MG
Methyl β-D-mannopyranoside isopropylate ≥98%
Methyl β-D-mannopyranoside isopropylate
Formula: C₇H₁₄O₆·(CH₃)₂CHOH
MW: 254,277 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03453195
CAS Number: 911673-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR