Propargyl α-D-galactopyranoside ≥98%

Supplier: Apollo Scientific

Synonyms: 2-Propynyl α-D-galactopyranoside, 2-Propyn-1-yl α-D-galactoside

BICL4265-100MG BICL4265-250MG BICL4265-500MG
APOSBICL4265-100MGEA 405 EUR
APOSBICL4265-100MG APOSBICL4265-250MG APOSBICL4265-500MG
Propargyl α-D-galactopyranoside ≥98%
Propargyl α-D-galactopyranoside
Formula: C₉H₁₄O₆
Storage Temperature: Freezer
MDL Number: MFCD15145085
CAS Number: 913074-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR