2,3,6-Tri-O-(carboxymethyl)-D-glucose ≥95%

Supplier: Apollo Scientific

BICL4291-1MG BICL4291-2MG
APOSBICL4291-1MGEA 768 EUR
APOSBICL4291-1MG APOSBICL4291-2MG
2,3,6-Tri-O-(carboxymethyl)-D-glucose ≥95%
2,3,6-Tri-O-(carboxymethyl)-D-glucose
Storage Temperature: Ambient
CAS Number: 108844-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR