4-Methylphenyl 4,6-O-benzylidene-1-thio-β-D-glucopyranoside ≥98%

Supplier: Apollo Scientific

Synonyms: Tolyl 4,6-O-benzylidene-1-thio-β-D-glucopyranoside, Thiocresyl 4,6-O-benzylidene-β-D-glucopyranoside

BICL4352-1G BICL4352-5G BICL4352-2G
APOSBICL4352-1GEA 337 EUR
APOSBICL4352-1G APOSBICL4352-5G APOSBICL4352-2G
4-Methylphenyl 4,6-O-benzylidene-1-thio-β-D-glucopyranoside ≥98%
4-Methylphenyl 4,6-O-benzylidene-1-thio-β-D-glucopyranoside
Formula: C₂₀H₂₂O₅S
MW: 374,451 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13182352
CAS Number: 868241-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR