5-Bromo-4-chloro-3-indolyl β-D-xylopyranoside

Supplier: Apollo Scientific
Warning

Synonyms: X-xyloside

BICS0109-100MG BICS0109-25MG
APOSBICS0109-100MGEA 143 EUR
APOSBICS0109-100MG APOSBICS0109-25MG
5-Bromo-4-chloro-3-indolyl β-D-xylopyranoside
5-Bromo-4-chloro-3-indolyl β-D-xylopyranoside
Formula: C₁₃H₁₃BrClNO₅
Storage Temperature: Ambient
MDL Number: MFCD00152013
CAS Number: 207606-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR