Fusarenon X

Supplier: Apollo Scientific
Danger

Synonyms: 12,13-epoxy-3 α,4 β,7 β,15-tetrahydroxytrichothec-9-en-8-one 4-acetate, Nivalenol monoacetate

BIF1007-1MG BIF1007-5MG
APOSBIF1007-1MGEA 364 EUR
APOSBIF1007-1MG APOSBIF1007-5MG
Fusarenon X
Fusarenon X
Formula: C₁₇H₂₂O₈
MW: 354,352 g/mol
Melting Pt: 91…92 °C
Storage Temperature: Freezer
MDL Number: MFCD00078778
CAS Number: 23255-69-8
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR