4-Methylumbelliferyl β-D-mannopyranoside

Supplier: Apollo Scientific
Warning

BIM2045-100MG BIM2045-50MG
APOSBIM2045-100MGEA 132 EUR
APOSBIM2045-100MG APOSBIM2045-50MG
4-Methylumbelliferyl β-D-mannopyranoside
4-Methylumbelliferyl β-D-mannopyranoside
Formula: C₁₆H₁₈O₈
MW: 338,309 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00069855
CAS Number: 67909-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR