Perillartine 95%

Supplier: Apollo Scientific
Warning

Synonyms: (NZ)-N-[(4-prop-1-en-2-ylcyclohexen-1-yl)methylidene]hydroxylamine, (E)-4-(Prop-1-en-2-yl)cyclohex-1-enecarbaldehyde oxime

BIP4001-100G BIP4001-25G
APOSBIP4001-100GEA 498 EUR
APOSBIP4001-100G APOSBIP4001-25G
Perillartine 95%
Perillartine
Formula: C₁₀H₁₅NO
MW: 165,232 g/mol
Melting Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00019421
CAS Number: 30950-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR