Fmoc-N-amido-PEG9-acid

Supplier: Apollo Scientific

Synonyms: Fmoc-NH-PEG9-CH2CH2COOH, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28,31-decaoxa-4-azatetratriacontan-34-oic acid

BIPG1403-100MG
APOSBIPG1403-100MGEA 30.6 EUR
APOSBIPG1403-100MG
Fmoc-N-amido-PEG9-acid
Fmoc-N-amido-PEG9-acid
Formula: C₃₆H₅₃NO₁₃
MW: 707,805 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28385470
CAS Number: 1191064-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR