Hydroxy-PEG2-acid

Supplier: Apollo Scientific

Synonyms: 3-(2-(2-Hydroxyethoxy)ethoxy)propanoic acid

BIPG1428-250MG BIPG1428-1G BIPG1428-500MG
APOSBIPG1428-250MGEA 139 EUR
APOSBIPG1428-250MG APOSBIPG1428-1G APOSBIPG1428-500MG
Hydroxy-PEG2-acid
Hydroxy-PEG2-acid
Formula: C₇H₁₄O₅
MW: 178,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20646030
CAS Number: 1334286-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR