Umckalin
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
7-Hydroxy-5,6-dimethoxy-2H-1-benzopyran-2-one, 5-Methoxyscopoletin, 7-Hydroxy-5,6-dimethoxycoumarin
BIU1200-10MG
BIU1200-5MG
APOSBIU1200-10MGEA
741
EUR
APOSBIU1200-10MG
APOSBIU1200-5MG
Umckalin
Umckalin
Limited shelf life
Formula:
C₁₁H₁₀O₅ MW: 222,194 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16036238 CAS Number: 43053-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...