(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: (2S)-2-(3,4,5-Trimethoxyphenyl)butyric acid

OR01538-1G OR01538-250MG OR01538-5G
APOSOR01538-1GEA 133 EUR
APOSOR01538-1G APOSOR01538-250MG APOSOR01538-5G
(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid 98%
(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid
Formula: C₁₃H₁₈O₅
MW: 254,279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06245370
CAS Number: 195202-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR