4-Bromophenylhydrazine

Supplier: Apollo Scientific
Warning

OR016911-5G OR016911-1G
APOSOR016911-5GEA 0 EUR
APOSOR016911-5G APOSOR016911-1G
4-Bromophenylhydrazine
4-Bromophenylhydrazine
Formula: C₆H₇BrN₂
Melting Pt: 102…104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01935685
CAS Number: 589-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR