4-(3-Bromophenyl)-1H-pyrazole 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(1H-Pyrazol-4-yl)bromobenzene
OR0211-1G
OR0211-5G
OR0211-10G
APOSOR0211-1GEA
238
EUR
APOSOR0211-1G
APOSOR0211-5G
APOSOR0211-10G
4-(3-Bromophenyl)-1H-pyrazole 95+%
4-(3-Bromophenyl)-1H-pyrazole
Formula:
C₉H₇BrN₂ MW: 223,069 g/mol Melting Pt: 140…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD06245520 CAS Number: 916792-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...