2-[(Piperazin-1-yl)methyl]benzonitrile 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(2-Cyanobenzyl)piperazine, 1-[(2-Cyanophenyl)methyl]piperazine
OR0228-1G
OR0228-250MG
OR0228-5G
APOSOR0228-1GEA
129
EUR
APOSOR0228-1G
APOSOR0228-250MG
APOSOR0228-5G
2-[(Piperazin-1-yl)methyl]benzonitrile 97%
2-[(Piperazin-1-yl)methyl]benzonitrile
Formula:
C₁₂H₁₅N₃ MW: 201,268 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04972582 CAS Number: 174609-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...