2-(Piperazin-1-ylcarbonyl)benzoic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Phtaloylpiperazine
OR0823-10G
OR0823-2G
APOSOR0823-10GEA
369
EUR
APOSOR0823-10G
APOSOR0823-2G
2-(Piperazin-1-ylcarbonyl)benzoic acid
2-(Piperazin-1-ylcarbonyl)benzoic acid
Formula:
C₁₂H₁₄N₂O₃ MW: 234,251 g/mol Melting Pt: >245 °C Storage Temperature: Ambient |
MDL Number:
MFCD03042136 CAS Number: 37618-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...