2-(Piperazin-1-yl)-1,3,5-triazine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(1,3,5-Triazin-2-yl)piperazine, 2-(Piperazin-1-yl)-s-triazine
OR0838-250MG
OR0838-50MG
APOSOR0838-250MGEA
130
EUR
APOSOR0838-250MG
APOSOR0838-50MG
2-(Piperazin-1-yl)-1,3,5-triazine
2-(Piperazin-1-yl)-1,3,5-triazine
Formula:
C₇H₁₁N₅ MW: 165,196 g/mol Melting Pt: 78…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973348 CAS Number: 59215-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...