1-[2-(1H-Imidazol-1-yl)ethyl]piperazine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-[2-(Piperazin-1-yl)ethyl]-1H-imidazole
OR0883-10G
OR0883-1G
OR0883-2G
APOSOR0883-10GEA
389
EUR
APOSOR0883-10G
APOSOR0883-1G
APOSOR0883-2G
1-[2-(1H-Imidazol-1-yl)ethyl]piperazine
1-[2-(1H-Imidazol-1-yl)ethyl]piperazine
Formula:
C₉H₁₆N₄ MW: 180,25 g/mol Boiling Pt: 124…126 °C (0,3 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD03412120 CAS Number: 381721-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...