N-(2-Aminoethyl)-5-chloroisoquinoline-8-sulphonamide

Supplier: Apollo Scientific
Danger

OR0885T-10MG OR0885T-25MG OR0885T-5MG
APOSOR0885T-10MGEA 400 EUR
APOSOR0885T-10MG APOSOR0885T-25MG APOSOR0885T-5MG
N-(2-Aminoethyl)-5-chloroisoquinoline-8-sulphonamide
N-(2-Aminoethyl)-5-chloroisoquinoline-8-sulphonamide

CKI-7 has potent inhibitory action against casein kinase 1 with respect to ATP binding and a much weaker effect on casein kinase 2 and other protein kinases.

Formula: C₁₁H₁₂ClN₃O₂S
MW: 285,75 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00269886
CAS Number: 120615-25-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR