Ethyl 3-(piperazin-1-yl)propanoate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(3-Ethoxy-3-oxoprop-1-yl)piperazine
OR0949-1G
OR0949-5G
APOSOR0949-1GEA
284
EUR
APOSOR0949-1G
APOSOR0949-5G
Ethyl 3-(piperazin-1-yl)propanoate
Ethyl 3-(piperazin-1-yl)propanoate
Formula:
C₉H₁₈N₂O₂ MW: 186,251 g/mol Boiling Pt: 89…93 °C (0,1 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD03410256 CAS Number: 43032-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...