3-(Prop-2-yn-1-ylcarbamoyl)acrylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 4-Oxo-4-(prop-2-yn-1-ylamino)but-2-enoic acid

OR10264-1G OR10264-250MG
APOSOR10264-1GEA 139 EUR
APOSOR10264-1G APOSOR10264-250MG
3-(Prop-2-yn-1-ylcarbamoyl)acrylic acid
3-(Prop-2-yn-1-ylcarbamoyl)acrylic acid
Formula: C₇H₇NO₃
MW: 153,135 g/mol
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00111718
CAS Number: 671188-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR