7-Aminothieno[2,3-b]pyrazine-6-carboxylic acid

Supplier: Apollo Scientific
Warning

OR103648-1G OR103648-5G
APOSOR103648-1GEA 125 EUR
APOSOR103648-1G APOSOR103648-5G
7-Aminothieno[2,3-b]pyrazine-6-carboxylic acid
7-Aminothieno[2,3-b]pyrazine-6-carboxylic acid
Formula: C₇H₅N₃O₂S
MW: 195,199 g/mol
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD08271964
CAS Number: 56881-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR