Diethyl (3-chloro-2-oxoprop-1-yl)phosphonate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Chloro-3-(diethoxyphosphoryl)acetone, Diethyl [(chloroacetyl)methyl]phosphonate
OR10698-1G
OR10698-250MG
APOSOR10698-1GEA
986
EUR
APOSOR10698-1G
APOSOR10698-250MG
Diethyl (3-chloro-2-oxoprop-1-yl)phosphonate
Diethyl (3-chloro-2-oxoprop-1-yl)phosphonate
Formula:
C₇H₁₄ClO₄P Boiling Pt: 94…95 °C (0,07 mm Hg) Density: 1,219 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD04038926 CAS Number: 67257-29-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...