4-Bromophenoxyacetaldehyde diethylacetal

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromo-4-(2,2-diethoxyethoxy)benzene

OR10835-1G OR10835-5G
APOSOR10835-1GEA 29 EUR
APOSOR10835-1G APOSOR10835-5G
4-Bromophenoxyacetaldehyde diethylacetal
4-Bromophenoxyacetaldehyde diethylacetal
Formula: C₁₂H₁₇BrO₃
MW: 289,166 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03411183
CAS Number: 112598-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR