4-Methoxy-1-methyl-1H-indazol-3-amine

Supplier: Apollo Scientific
Warning

OR110274-1G OR110274-250MG
APOSOR110274-1GEA 221 EUR
APOSOR110274-1G APOSOR110274-250MG
4-Methoxy-1-methyl-1H-indazol-3-amine
4-Methoxy-1-methyl-1H-indazol-3-amine
Formula: C₉H₁₁N₃O
MW: 177,203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176793
CAS Number: 1305711-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR