5-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,3-oxazole

Supplier: Apollo Scientific

OR110279-1G OR110279-250MG
APOSOR110279-1GEA 290 EUR
APOSOR110279-1G APOSOR110279-250MG
5-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,3-oxazole
5-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,3-oxazole
Formula: C₁₁H₉NO₃
MW: 203,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13191320
CAS Number: 1305712-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR