Methyl 2-acetyl-2H-indazole-5-carboxylate

Supplier: Apollo Scientific

OR110307-1G OR110307-250MG OR110307-5G
APOSOR110307-1GEA 231 EUR
APOSOR110307-1G APOSOR110307-250MG APOSOR110307-5G
Methyl 2-acetyl-2H-indazole-5-carboxylate
Methyl 2-acetyl-2H-indazole-5-carboxylate
Formula: C₁₁H₁₀N₂O₃
MW: 218,209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14635903
CAS Number: 1308649-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR