4-(Hydrazinocarbonyl)benzoic acid

Supplier: Apollo Scientific
Warning

OR110418-1G OR110418-250MG
APOSOR110418-1GEA 168 EUR
APOSOR110418-1G APOSOR110418-250MG
4-(Hydrazinocarbonyl)benzoic acid
4-(Hydrazinocarbonyl)benzoic acid
Formula: C₈H₈N₂O₃
MW: 180,161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041381
CAS Number: 46206-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR