2-Methyl-1-nitro-4-phenoxybenzene

Supplier: Apollo Scientific

OR110478-1G OR110478-5G
APOSOR110478-1GEA 87 EUR
APOSOR110478-1G APOSOR110478-5G
2-Methyl-1-nitro-4-phenoxybenzene
2-Methyl-1-nitro-4-phenoxybenzene
Formula: C₁₃H₁₁NO₃
MW: 229,231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01764677
CAS Number: 112880-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR