2-(4-Bromophenoxy)benzaldehyde

Supplier: Apollo Scientific

OR110489-1G OR110489-5G
APOSOR110489-1GEA 186 EUR
APOSOR110489-1G APOSOR110489-5G
2-(4-Bromophenoxy)benzaldehyde
2-(4-Bromophenoxy)benzaldehyde
Formula: C₁₃H₉BrO₂
MW: 277,113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11156483
CAS Number: 364323-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR