[2-(4-Methylphenoxy)phenyl]methanol

Supplier: Apollo Scientific

OR110504-1G OR110504-5G
APOSOR110504-1GEA 248 EUR
APOSOR110504-1G APOSOR110504-5G
[2-(4-Methylphenoxy)phenyl]methanol
[2-(4-Methylphenoxy)phenyl]methanol
Formula: C₁₄H₁₄O₂
MW: 214,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02102517
CAS Number: 25562-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR