4-[2-(4-tert-Butoxycarbonylpiperazin-1-yl)ethoxy]acetophenone

Supplier: Apollo Scientific

OR110598-1G OR110598-10G OR110598-5G
APOSOR110598-1GEA 248 EUR
APOSOR110598-1G APOSOR110598-10G APOSOR110598-5G
4-[2-(4-tert-Butoxycarbonylpiperazin-1-yl)ethoxy]acetophenone
4-[2-(4-tert-Butoxycarbonylpiperazin-1-yl)ethoxy]acetophenone
Formula: C₁₉H₂₈N₂O₄
MW: 348,437 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756240
CAS Number: 1227954-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR