(4,5,6,7-Tetrahydro-1-benzothiophen-2-yl)methanol

Supplier: Apollo Scientific
Warning

OR110614-1G OR110614-5G OR110614-250MG
APOSOR110614-1GEA 263 EUR
APOSOR110614-1G APOSOR110614-5G APOSOR110614-250MG
(4,5,6,7-Tetrahydro-1-benzothiophen-2-yl)methanol
(4,5,6,7-Tetrahydro-1-benzothiophen-2-yl)methanol
Formula: C₉H₁₂OS
MW: 168,256 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059489
CAS Number: 51632-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR