4,5,6,7-Tetrahydro-1-benzothiophene-2-carbaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR110615-1G
OR110615-250MG
APOSOR110615-1GEA
207
EUR
APOSOR110615-1G
APOSOR110615-250MG
4,5,6,7-Tetrahydro-1-benzothiophene-2-carbaldehyde
4,5,6,7-Tetrahydro-1-benzothiophene-2-carbaldehyde
Formula:
C₉H₁₀OS MW: 166,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16631271 CAS Number: 57021-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...