6-Hydroxy-2,2-dimethyl-2,3-dihydro-4H-chromen-4-one
Supplier: Apollo Scientific
|
Warning
|
OR110738-1G
OR110738-5G
APOSOR110738-1GEA
14.7
EUR
APOSOR110738-1G
APOSOR110738-5G
6-Hydroxy-2,2-dimethyl-2,3-dihydro-4H-chromen-4-one
6-Hydroxy-2,2-dimethyl-2,3-dihydro-4H-chromen-4-one
Formula:
C₁₁H₁₂O₃ MW: 192,211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03830209 CAS Number: 31366-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...