1-(3,4-Dichlorobenzyl)homopiperazine
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-(3,4-Dichlorobenzyl)-1,4-diazepane
OR110776-1G
OR110776-5G
OR110776-10G
APOSOR110776-1GEA
149
EUR
APOSOR110776-1G
APOSOR110776-5G
APOSOR110776-10G
1-(3,4-Dichlorobenzyl)homopiperazine
1-(3,4-Dichlorobenzyl)homopiperazine
Formula:
C₁₂H₁₆Cl₂N₂ MW: 259,175 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01311955 CAS Number: 78251-53-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...