2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
Supplier: Apollo Scientific
OR110882-1G
OR110882-5G
APOSOR110882-1GEA
81.5
EUR
APOSOR110882-1G
APOSOR110882-5G
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
Formula:
C₁₂H₁₃NO₃ MW: 219,237 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04353248 CAS Number: 950252-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...