N'-Hydroxy-1-methyl-1H-indole-4-carboximidamide

Supplier: Apollo Scientific
Warning

OR110894-1G OR110894-5G
APOSOR110894-1GEA 14.7 EUR
APOSOR110894-1G APOSOR110894-5G
N'-Hydroxy-1-methyl-1H-indole-4-carboximidamide
N'-Hydroxy-1-methyl-1H-indole-4-carboximidamide
Formula: C₁₀H₁₁N₃O
Storage Temperature: Ambient
MDL Number: MFCD27995617
CAS Number: 1597528-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR