Ethyl 7-bromo-5-chlorobenzo[b]furan-2-carboxylate

Supplier: Apollo Scientific
Warning

OR110908-1G OR110908-5G
APOSOR110908-1GEA 192 EUR
APOSOR110908-1G APOSOR110908-5G
Ethyl 7-bromo-5-chlorobenzo[b]furan-2-carboxylate
Ethyl 7-bromo-5-chlorobenzo[b]furan-2-carboxylate
Formula: C₁₁H₈BrClO₃
MW: 303,536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28040507
CAS Number: 1823331-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR