1,3-Benzoxazole-5-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
OR110970-1G
OR110970-250MG
APOSOR110970-1GEA
234
EUR
APOSOR110970-1G
APOSOR110970-250MG
1,3-Benzoxazole-5-carbonitrile
1,3-Benzoxazole-5-carbonitrile
Formula:
C₈H₄N₂O MW: 144,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16817443 CAS Number: 132227-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...