3-(6-Methoxy-2-naphthyl)-1H-pyrazole
Supplier: Apollo Scientific
OR111017-1G
OR111017-250MG
APOSOR111017-1GEA
232
EUR
APOSOR111017-1G
APOSOR111017-250MG
3-(6-Methoxy-2-naphthyl)-1H-pyrazole
3-(6-Methoxy-2-naphthyl)-1H-pyrazole
Formula:
C₁₄H₁₂N₂O MW: 224,258 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03407411 CAS Number: 258518-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...