Ethyl (2-acetyl-4-methylphenoxy)acetate
Supplier: Apollo Scientific
OR111019-1G
OR111019-5G
APOSOR111019-1GEA
198
EUR
APOSOR111019-1G
APOSOR111019-5G
Ethyl (2-acetyl-4-methylphenoxy)acetate
Ethyl (2-acetyl-4-methylphenoxy)acetate
Formula:
C₁₃H₁₆O₄ Storage Temperature: Ambient |
MDL Number:
MFCD12417043 CAS Number: 58335-85-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...