Ethyl 2,5-dichlorothiophene-3-carboxylate

Supplier: Apollo Scientific
Warning

OR111044-1G OR111044-250MG
APOSOR111044-1GEA 275 EUR
APOSOR111044-1G APOSOR111044-250MG
Ethyl 2,5-dichlorothiophene-3-carboxylate
Ethyl 2,5-dichlorothiophene-3-carboxylate
Formula: C₇H₆Cl₂O₂S
MW: 225,092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06205024
CAS Number: 130562-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR